Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166608.1 | NP_001160080.1 | 11 - 12 | 1930 | 101 | |
NM_001166609.1 | NP_001160081.1 | 11 - 12 | 1815 | 101 | ||
NM_001166610.1 | NP_001160082.1 | 11 - 12 | 1682 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Probe spans exons |