Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166611.1 | NP_001160083.1 | 2 - 3 | 263 | 71 | |
GenBank mRNA | BC148137.1 | - | 2 - 3 | 212 | 71 | |
EV659412.1 | - | 2 - 3 | 272 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |