Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167896.2 | NP_001161368.1 | 2 - 3 | 641 | 79 | |
GenBank mRNA | BC105241.1 | - | 2 - 3 | 332 | 79 | |
BC150062.1 | - | 2 - 3 | 640 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |