Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191151.1 | NP_001178080.1 | 2 - 3 | 2751 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |