Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206822.1 | NP_001193751.1 | 19 - 20 | 3782 | 86 | |
GenBank mRNA | EH158845.1 | - | 1 - 2 | 385 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |