Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206822.1 | NP_001193751.1 | 7 - 8 | 874 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |