Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206862.1 | NP_001193791.1 | 2 - 3 | 322 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |