Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206869.2 | NP_001193798.2 | 2 - 3 | 523 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |