Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206872.1 | NP_001193801.1 | 3 - 4 | 969 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |