Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGLON5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206875.1 | NP_001193804.1 | 1 - 2 | 82 | 83 | |
GenBank mRNA | EH200051.1 | - | 1 - 2 | 60 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |