Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206899.2 | NP_001193828.1 | 4 - 5 | 798 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |