Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191444.2 | NP_001178373.1 | 5 - 6 | 1491 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |