Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244204.1 | NP_001231133.1 | 3 - 4 | 461 | 101 | |
GenBank mRNA | BC116160.1 | - | 3 - 4 | 450 | 101 | |
EE251296.1 | - | 3 - 4 | 462 | 101 | ||
EV744049.1 | - | 5 - 6 | 654 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |