Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244318.2 | NP_001231247.1 | 8 - 9 | 927 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |