Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245925.1 | NP_001232854.1 | 1 - 2 | 183 | 74 | |
GenBank mRNA | HM036582.1 | - | 1 - 2 | 183 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |