Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173957.2 | NP_776382.1 | 4 - 5 | 664 | 78 | |
GenBank mRNA | AB035446.1 | - | 4 - 5 | 664 | 78 | |
BC103461.1 | - | 4 - 5 | 676 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |