Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173957.2 | NP_776382.1 | 5 - 6 | 794 | 89 | |
GenBank mRNA | AB035446.1 | - | 5 - 6 | 794 | 89 | |
BC103461.1 | - | 5 - 6 | 806 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |