Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174568.3 | NP_776993.1 | 2 - 3 | 897 | 83 | |
GenBank mRNA | AJ401269.2 | - | 2 - 3 | 383 | 83 | |
BC102365.1 | - | 2 - 3 | 887 | 83 | ||
EV617074.1 | - | 1 - 2 | 156 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |