Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THUMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192222.3 | NP_001179151.2 | 9 - 10 | 1198 | 63 | |
GenBank mRNA | BC149576.1 | - | 9 - 10 | 901 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |