Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192266.1 | NP_001179195.1 | 4 - 5 | 481 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |