Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDR2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192968.1 | NP_001179897.1 | 2 - 3 | 192 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |