Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193009.2 | NP_001179938.1 | 3 - 4 | 486 | 61 | |
GenBank mRNA | EH149263.1 | - | 1 - 2 | 514 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |