Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193117.1 | NP_001180046.1 | 2 - 3 | 305 | 103 | |
GenBank mRNA | FE015823.1 | - | 2 - 3 | 305 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |