Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193152.2 | NP_001180081.1 | 2 - 3 | 348 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |