Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193253.1 | NP_001180182.1 | 10 - 11 | 2292 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |