Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTSSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195042.1 | NP_001181971.1 | 1 - 2 | 127 | 78 | |
GenBank mRNA | BC134492.1 | - | 1 - 2 | 127 | 78 | |
BT021042.1 | - | 1 - 2 | 131 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |