Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASMTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035058.1 | NP_001030230.1 | 4 - 5 | 370 | 68 | |
GenBank mRNA | BC102999.1 | - | 4 - 5 | 370 | 68 | |
BT026261.1 | - | 4 - 5 | 394 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |