Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199005.3 | NP_001185934.1 | 2 - 3 | 256 | 98 | |
NM_001199006.2 | NP_001185935.1 | 2 - 3 | 256 | 98 | ||
GenBank mRNA | BC151678.1 | - | 2 - 3 | 256 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |