Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14H8orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199131.1 | NP_001186060.1 | 1 - 2 | 540 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |