Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GID4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199762.1 | NP_001186691.1 | 4 - 5 | 666 | 81 | |
GenBank mRNA | BC102715.1 | - | 4 - 5 | 530 | 81 | |
DT807444.1 | - | 4 - 5 | 449 | 81 | ||
DT829639.1 | - | 4 - 5 | 530 | 81 | ||
EE341469.1 | - | 2 - 3 | 142 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |