Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TASOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205393.2 | NP_001192322.1 | 10 - 11 | 1258 | 98 | |
GenBank mRNA | BC151347.1 | - | 2 - 3 | 136 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |