Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270317.4 | NP_001257246.2 | 19 - 20 | 5003 | 147 | |
NM_001373583.1 | NP_001360775.1 | 19 - 20 | 4970 | 147 | ||
NM_001381151.1 | NP_001368758.1 | 19 - 20 | 4986 | 147 | ||
NM_001381152.1 | NP_001368759.1 | 19 - 20 | 4986 | 147 | ||
NM_001381153.1 | NP_001368756.1 | 19 - 20 | 4953 | 147 | ||
NM_001381154.1 | NP_001368757.1 | 19 - 20 | 4953 | 147 | ||
NM_001381155.1 | NP_001368146.1 | 19 - 20 | 4980 | 147 | ||
NM_001381156.2 | NP_001368760.1 | 19 - 20 | 4976 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |