Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322663.4 | NP_001309674.1 | 14 - 15 | 1959 | 72 | |
NM_001322664.3 | NP_001309467.1 | 13 - 14 | 1920 | 72 | ||
NM_001322665.4 | NP_001309678.1 | 9 - 10 | 1031 | 72 | ||
NM_001322667.4 | NP_001309677.1 | 3 - 4 | 494 | 72 | ||
NM_001322668.4 | NP_001309676.1 | 3 - 4 | 407 | 72 | ||
NM_001383675.1 | 13 - 14 | 1913 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |