Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322663.4 | NP_001309674.1 | 16 - 17 | 2388 | 97 | |
NM_001322664.3 | NP_001309467.1 | 15 - 16 | 2349 | 97 | ||
NM_001322665.4 | NP_001309678.1 | 11 - 12 | 1460 | 97 | ||
NM_001322667.4 | NP_001309677.1 | 5 - 6 | 923 | 97 | ||
NM_001322668.4 | NP_001309676.1 | 5 - 6 | 836 | 97 | ||
NM_001383675.1 | 15 - 16 | 2342 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |