Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322663.4 | NP_001309674.1 | 21 - 22 | 3485 | 62 | |
NM_001322664.3 | NP_001309467.1 | 20 - 21 | 3446 | 62 | ||
NM_001322665.4 | NP_001309678.1 | 16 - 17 | 2557 | 62 | ||
NM_001322667.4 | NP_001309677.1 | 10 - 11 | 2020 | 62 | ||
NM_001322668.4 | NP_001309676.1 | 10 - 11 | 1933 | 62 | ||
NM_001322669.4 | NP_001309675.1 | 1 - 2 | 58 | 62 | ||
NM_001383675.1 | 20 - 21 | 3439 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |