Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078059.6 | NP_510460.2 | 3 - 4 | 524 | 84 | |
GenBank mRNA | AY064464.1 | - | 3 - 4 | 537 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |