Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pag-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078079.6 | NP_510480.1 | 5 - 6 | 623 | 69 | |
GenBank mRNA | U63996.1 | - | 5 - 6 | 621 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |