Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078107.6 | NP_510508.1 | 10 - 11 | 1574 | 113 | |
GenBank mRNA | AB012700.1 | - | 10 - 11 | 1596 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |