Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: svh-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270332.3 | NP_001257261.1 | 6 - 7 | 1223 | 72 | |
NM_001383683.1 | 7 - 8 | 1470 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |