Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270335.2 | NP_001257264.1 | 7 - 8 | 1337 | 112 | |
NM_001270336.3 | NP_001257265.1 | 6 - 7 | 1134 | 112 | ||
NM_001313436.5 | NP_001300365.1 | 8 - 9 | 1284 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |