Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: let-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078262.6 | NP_510663.1 | 9 - 10 | 936 | 80 | |
NM_078263.4 | NP_510664.1 | 9 - 10 | 951 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |