Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078280.8 | NP_510681.4 | 21 - 22 | 4719 | 77 | |
GenBank mRNA | DQ360811.1 | - | 21 - 22 | 4003 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |