Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sod-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078363.8 | NP_510764.1 | 3 - 4 | 420 | 88 | |
GenBank mRNA | X85790.1 | - | 380 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |