Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tps-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047837.4 | NP_001041302.1 | 14 - 15 | 2678 | 73 | |
NM_001047838.4 | NP_001041303.1 | 13 - 14 | 2510 | 73 | ||
GenBank mRNA | AJ512332.1 | - | 14 - 15 | 2621 | 73 | |
AJ811573.1 | - | 14 - 15 | 2649 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |