Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tps-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047837.4 | NP_001041302.1 | 9 - 10 | 1408 | 117 | |
NM_001047838.4 | NP_001041303.1 | 8 - 9 | 1240 | 117 | ||
GenBank mRNA | AJ512332.1 | - | 9 - 10 | 1351 | 117 | |
AJ811573.1 | - | 9 - 10 | 1379 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |