Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029682.5 | NP_001024853.1 | 10 - 11 | 1154 | 84 | |
NM_001029684.6 | NP_001024855.1 | 9 - 10 | 1036 | 84 | ||
GenBank mRNA | AY204163.1 | - | 9 - 10 | 1020 | 84 | |
AY204164.1 | - | 9 - 10 | 1034 | 84 | ||
AY204165.1 | - | 9 - 10 | 1049 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |