Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nlp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270358.4 | NP_001257287.1 | 1 - 2 | 112 | 77 | |
NM_001270359.4 | NP_001257288.1 | 1 - 2 | 100 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |