Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306422.3 | NP_001293351.1 | 7 - 8 | 1633 | 94 | |
NM_170819.6 | NP_740802.1 | 9 - 10 | 2030 | 94 | ||
NR_131352.1 | 9 - 10 | 1869 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Caenorhabditis elegans Probe spans exons |