Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpxl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170821.6 | NP_740805.1 | 4 - 5 | 1427 | 78 | |
GenBank mRNA | DQ178245.1 | - | 4 - 5 | 1427 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |