Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aman-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374889.1 | NP_001361919.1 | 11 - 12 | 1547 | 79 | |
NM_001374890.2 | NP_001362173.1 | 11 - 12 | 1554 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |